missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MafG Protein

Highly purified. Generating reliable and reproducible results.

Marca:  Novus Biologicals™ NBP1-72507-250UG

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216012

  • 1361.74€ / 250 microgramos
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-162 of Human MafG The Recombinant Human MafG Protein is derived from E. coli. The Recombinant Human MafG Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

SDS-PAGE
250μg
Store at -80°C. Avoid freeze-thaw cycles.
MafG
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 40% glycerol, 0.15M NaCl, 1mM DTT
Human
RUO
Unconjugated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales