missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human RUNX3/CBFA3 His Protein

Highly purified. Generating reliable and reproducible results.

Marca:  Novus Biologicals™ NBP1-44462

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215222

  • 384.44€ / 0.05 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 53-186 of Human RUNX3/CBFA3 The Recombinant Human RUNX3/CBFA3 Protein is derived from E. coli. The Recombinant Human RUNX3/CBFA3 Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

NP_004341
20mM Tris-HCl buffer (pH8.0), 20% glycerol, 0.1 M NaCl, 1mM DTT
17.1kDa
Protein
−80°C. Avoid freeze-thaw cycles.
>95%, by SDS-PAGE
ELISA, SDS-PAGE
864
RUNX3/CBFA3 Protein
MGSSHHHHHH SSGLVPRGSH MRSMVDVLAD HAGELVRTDS PNFLCSVLPS HWRCNKTLPV AFKVVALGDV PDGTVVTVMA GNDENYSAEL RNASAVMKNQ VARFNDLRFV GRSGRGKSFT LTITVFTNPT QVATYHRAIK VTVDGPREPR RHRQK
Human
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales