missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PA2G4/EBP1 His Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Marca:  Novus Biologicals™ NBP1-44474

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214421

  • 384.44€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-394 of Human PA2G4/EBP1 The Recombinant Human PA2G4/EBP1 Protein is derived from E. coli. The Recombinant Human PA2G4/EBP1 Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

Q9UQ80
20mM Tris-HCl buffer (pH8.0), 10% glycerol
44.8kDa
Protein
E. Coli
−80°C. Avoid freeze-thaw cycles.
Unconjugated
Recombinant
SDS-PAGE
5036
PA2G4/EBP1 Protein
0.1mg
PA2G4, 1-394aa, Human, His tag, E.coli (NP_006182).
RUO
Human
>95%, by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales