Learn More
Invitrogen™ Novex™ Western-Star™ Immunodetection System
High sensitivity, high signal intensity, chemiluminescence-based immunodetection system
Marca:  Invitrogen™ T1048
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12664295
  • 540.00€ / cada uno
Fecha de envío estimada
del proveedor 03-02-2020
Añadir a la cesta
Especificaciones
Especificaciones
Western-SuperStar™ Reagent Set (containing Blocker, Wash Buffer, Assay Buffer, Substrate, Enhancer, Blot-Loc™ bags, Development Folders and pair of Tweezers), anti-mouse secondary antibody conjugated to alkaline phosphatase
AP (Alkaline Phosphatase) Substrate
30 Blots
Rabbit
Protein
Videos
Documentos
For Research Use Only. Not for use in diagnostic procedures.