missing translation for 'onlineSavingsMsg'
Learn More

Nova2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-85394-0.1ML

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214239

  • 455.71€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Nova2 Polyclonal specifically detects Nova2 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

Nova2
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
4858
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
neuro-oncological ventral antigen 2ANOVANOVA3Astrocytic NOVA1-like RNA-binding protein, neuro-oncological ventral antigen 3, RNA-binding protein Nova-2
The immunogen is a synthetic peptide directed towards the middle region of human Nova2
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales