missing translation for 'onlineSavingsMsg'
Learn More

Notch-1 Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Take advantage of a quality product that delivers accurate results with the Epredia™ Notch-1 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1339-P

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12669606

  • 540.00€ / 1 ml

Envío estimado: 02-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals, which include notch-1. notch-2, notch-3, and notch-4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins δ and Serrate have been identified as ligands of notch-1. Mature notch proteins are heterodimeric receptors derived from the cleavage of notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm). Alteration of notch-1 signaling or expression may contribute to tumorigenesis.

Host Species: Mouse

Clone: A6

Isotype: IgG2b

Species Reactivity: Human and Mouse. Others not known.

Epitope: Ligand binding region

Immunogen: Recombinant protein encoding the ligand binding region of human Notch-1

Molecular Weight: 290kDa

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Not verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Notch-1 Ab-1
Monoclonal
200μg/mL
Concentrated
Recombinant protein encoding the ligand binding region of human Notch-1
1mL
Neuroscience
Primary
Human, Murine
Flow Cytometry
A6
Unconjugated
Murine
IgG2b
RUO
Primary
Monoclonal
Videos
SDS
Documentos
Certificados