Learn More
R&D Systems™ Noradrenaline bitartrate
Product Code. 18707486
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
18707486 50 mg 50mg
1 options
This item is not returnable. View return policy
Product Code. 18707486 Supplier R&D Systems™ Supplier No. 5169/50
  • ONLINE EXCLUSIVE PRICE
    115.04€
    102.90€ / 50 miligramos
    Ahorro 12.14€
    10% Descuento
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Endogenous adrenergic hormone and neurotransmitter
Noradrenaline bitartrate is an endogenous adrenergic hormone and neurotransmitter. A classical stress hormone; underlies the fight or flight response, along with adrenaline. Increases heart rate. Vasoconstrictor.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mg
Product Line Tocris
Product Type Noradrenaline bitartrate
Purity >99%
Molecular Weight (g/mol) 319.26
Inhibitors Noradrenaline bitartrate
For Research Use Only