missing translation for 'onlineSavingsMsg'
Learn More

NOL12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15456-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814644

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a GTPase-activating protein (GAP) that associates with the Golgi apparatus and regulates the early secretory pathway of proteins. The encoded protein promotes hydrolysis of ADP-ribosylation factor 1 (ARF1)-bound GTP, which is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is a prerequisite for the fusion of these vesicles with target compartments. The activity of this protein is sensitive to phospholipids. Multiple transcript variants encoding different isoforms have been found for this gene. This gene was originally known as ARFGAP1, but that is now the name of a related but different gene.
Especificaciones

Especificaciones

NOL12
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5D1Z3, Q8BG17, Q9UGY1
NOL12, Nop25, nucleolar protein 12
Rabbit
IgG
20 μL
-20°C
Primary
362955, 79159, 97961
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
Unconjugated
NOL12
Liquid
Nol12
NOL12 Fusion Protein Ag7723
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Videos
SDS
Documentos
Certificados