missing translation for 'onlineSavingsMsg'
Learn More

NOB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10091-2-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816253

  • 109.00€ / 20 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5. 8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.
Especificaciones

Especificaciones

NOB1
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6VEU1, Q8BW10, Q9ULX3
ART 4, ART4, MST158, MSTP158, NOB1, NOB1P, Protein ART 4, PSMD8BP1, RNA binding protein NOB1
Rabbit
IgG
20 μL
-20°C
Primary
28987, 291996, 67619
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
NOB1
Liquid
NOB1
NOB1 Fusion Protein Ag0141
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados