missing translation for 'onlineSavingsMsg'
Learn More

NMDAR2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19953-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16819434

  • 109.00€ / 20 microlitros

Envío estimado: 20-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The antibody recognize the C-term of GRIN2A.

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Especificaciones

Especificaciones

NMDAR2A
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35436, Q12879
GluN2A, GRIN2A, hNR2A, NMDAR2A, NR2A
Rabbit
IgG
20 μL
-20°C
Primary
14811, 2903
Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
GRIN2A
Liquid
Grin2a
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados