missing translation for 'onlineSavingsMsg'
Learn More

NKX2.2 Antibody (rNX2/294), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-54345G

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214252

  • 538.86€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

NKX2.2 Monoclonal specifically detects NKX2.2 in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

NKX2.2
Monoclonal
DyLight 488
50 mM sodium borate with 0.05% sodium azide
NKX2-2
Recombinant full-length human NKX2.2 protein (Uniprot: O95096)
0.1 mL
4821.0
Store at 4°C in the dark.
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
rNX2/294
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B
Mouse
Protein A or G purified
Primary
Human, Mouse, Rat, Chicken
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales