missing translation for 'onlineSavingsMsg'
Learn More

NKX2.2 Antibody (NX2/1523), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-54328AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215183

  • 538.86€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

NKX2.2 Monoclonal specifically detects NKX2.2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

NKX2.2
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
NKX2-2
Recombinant fragment (around aa1-119) of human NKX2.2 protein (exact sequence is proprietary) (Uniprot: O95096)
Primary
Human
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
NX2/1523
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B
Mouse
0.1 mL
4821
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales