missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NKIRAS2 Recombinant Protein
Marca: Abnova™ H00028511-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH07450 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
46.75 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MGKSCKVVVCGQASVGKTSILEQLLYGNHVVGSEMIETQEDIYVGSIETDRGVREQVRFYDTRGLRDGAELPRHCFSCTDGYVLVYSTDSRESFQRVELLKKEIDKSKDKKEVTIVVLGNKCDLQEQRRVDPDVAQHWAKSEKVKLWEVSVADRRSLLEPFVYLASKMTQPQSKSAFPLSRKNKGSGSLDG | |
DKFZp434N1526/KBRAS2/MGC74742/kappaB-Ras2 | |
NKIRAS2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
28511 | |
NKIRAS2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NKIRAS2 | |
Human | |
Recombinant | |
Solution |