missing translation for 'onlineSavingsMsg'
Learn More

NKIRAS2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10174-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816383

  • 319.00€ / 150 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.
Especificaciones

Especificaciones

NKIRAS2
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NYR9
Kappa B Ras protein 2, kappaB Ras2, KBRAS2, NKIRAS2
Rabbit
IgG
150 μL
-20°C
Primary
28511
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
NKIRAS2
Liquid
NKIRAS2
NKIRAS2 Fusion Protein Ag0221
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados