missing translation for 'onlineSavingsMsg'
Learn More

NK-1.1 Mouse anti-Mouse, PerCP-Cy5.5, Clone: PK136, BD

Mouse Monoclonal Antibody

227.00€ - 535.00€

Especificaciones

Antígeno NK-1.1
Clon PK136
Concentración 0.2mg/mL
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15880779
Ver documentos Detalles de la promoción
BD Biosciences
561111
25 μg
227.00€
cada uno
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 23-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
10422345
Ver documentos Detalles de la promoción
BD Biosciences
551114
0.1 mg
535.00€
cada uno
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 23-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

In the mouse, at least three members of the Klrb (Killer cell lectin-like ; formerly NKR-P1) gene family have been identified (Klrb1a/NKR-P1A, Klrb1b/NKR-P1B, and Klrb1c/NKR-P1C); but in the human gene family, a single homologue has been designated KLRB1, NKR-P1A, or CD161. The KLRB1/NKR-P1 family of proteins are type-II-transmembrane C-type lectin receptors. KLRB1C/NKR-P1C activates NK-cell cytotoxicity, while KLRB1B/NKR-P1B functions as an inhibitory receptor. KLRB1B/NKR-P1B protein has intracellular Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM), while KLRB1C/NKR-P1C lacks ITIM and activates via association with Fc Receptor γchain. Strikingly, KLRB1B/NKR-P1B and KLRB1C/NKR-P1C share 96% amino acid sequence identity in their extracellular C-type lectin domains. The PK136 antibody reacts with the NK-1.1 surface antigen (CD161c) encoded by the Klrb1c/NKR-P1C gene expressed on natural killer (NK) cells in selected strains of mice (eg, C57BL, FVB/N, NZB, but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, 129) and the CD161b antigen encoded by the Klrb1b/NKR-P1B gene expressed only on Swiss NIH and SJL mice, but not on C57BL/6. Expression of KLRB1C/NKR-P1C protein is correlated with the ability to lyse tumor cells in vitro and to mediate rejection of bone marrow allografts. The NK-1.1 marker is useful in defining NK cells; however, the antigen is also expressed on a rare, specialized population of T lymphocytes (NK-T cells) and some cultured monocytes. Plate-bound PK136 mAb, in combination with low concentrations of IL-2, induces proliferation of a subset of NK cells.

Especificaciones

Especificaciones

NK-1.1
0.2mg/mL
Monoclonal
Mouse
Mouse
Mouse NK-1+ Spleen and Bone Marrow Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
PK136
Flow Cytometry
PerCP-Cyanine5.5
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Klrb1b, CD161b, Nkrp1b; Klrb1c, CD161c, NK1.1, Nkrp1c
Videos
SDS
Documentos

Documentos

Product Certifications

For Research Use Only.