Learn More
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves
Kimberly-Clark™ KLEENGUARD™ G20 Nitrile Gloves are an ambidextrous glove that provides protection against chemical splash in a wide range of applications.
Marca:  Kimberly-Clark™ 90092
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12648845
  • 313.00€ / paquete de 2500
Fecha estimada de envío
14-02-2020
Añadir a la cesta
Descripción
Descripción
  • PPE Category 3 protection
  • For increased protection against chemical splashes of several chemicals.
  • Provides exceptional touch sensitivity and textured fingertips to improve grip and the handling of small objects
  • AQL 0.65 for pinholes guarantees quality of production process, waste reduction and better protection
  • Independently accredited
  • Suitable for food handling
  • Beaded cuff for added strength and ease of donning
  • Latex Free
  • Powder free minimises srying effects of pwder and skin allergies
  • Ambidextrous
  • Portable dispenser box for mobility and easy access
Especificaciones
Especificaciones
Con reborde, largo
Chemical
2500/Pk.
Verde
Nitrilo
M
Nitrile Gloves
EN374-3: 2003, EN374-2:2003, EN388:2003