missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NINJ2 (Human) Recombinant Protein
Human NINJ2 full-length ORF ( AAH57766, 1 a.a. - 142 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00004815-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MESARENIDLQPGSSDPRSQPINLNHYATKKSVAESMLDVALFMSNAMRLKAVLEQGPSSHYYTTLVTLISLSLLLQVVIGVLLVVIARLNLNEVEKQWRLNQLNNAATILVFFTVVINVFITAFGAHKTGFLAARASRNPL
Especificaciones
AAH57766 | |
ninjurin 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NINJ2 | |
GST |
4815 | |
Wheat germ expression system | |
10 μg | |
FLJ56349 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |