missing translation for 'onlineSavingsMsg'
Learn More

Nicastrin Antibody (002), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89825UV

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215396

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

Nicastrin
Monoclonal
DyLight 350
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
2
Flow Cytometry
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin
This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (NP_056146.1; Met1-Glu669).
0.1 mL
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
23385
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales