Learn More
Thermo Scientific™ Human beta-NGF Recombinant Protein
Human beta-NGF Recombinant Protein
Marca: Thermo Scientific™ PHG0126
Detalles adicionales : Peso : 0.99000kg
Descripción
Human beta-NGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Recombinant human β-NGF is involved in both survival, differentiation, and function of various types of neurons.
Especificaciones
Cell Culture | |
Sequential Chromatography | |
Human | |
< 0.1 ng/μg | |
E. coli | |
Lyophilized | |
Gibco™ |
27 kDa | |
238 aa each (homodimer) | |
Carrier-Free | |
20 μg | |
Recombinant | |
Neurotrophic Factors | |
98 % |