missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFU1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25598-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that is localized to mitochondria and plays a critical role in iron-sulfur cluster biogenesis. The encoded protein assembles and transfers 4Fe-4S clusters to target apoproteins including succinate dehydrogenase and lipoic acid synthase. Mutations in this gene are a cause of multiple mitochondrial dysfunctions syndrome-1, and pseudogenes of this gene are located on the short arms of chromosomes 1 and 3. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Especificaciones
NFU1 | |
Polyclonal | |
Unconjugated | |
NFU1 | |
CGI 33, HIRA interacting protein 5, HIRIP5, Nfu, NFU1, NifU, NIFUC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27247, 297416, 56748 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QZ23, Q9UMS0 | |
Nfu1 | |
NFU1 Fusion Protein Ag22343 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |