missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NFE2 (Human) Recombinant Protein

Human NFE2 full-length ORF ( AAH05044, 1 a.a. - 373 a.a.) recombinant protein with GST-tag at N-terminal.

en la oferta

Especificaciones

Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ID de gen (Entrez) 4778
Ver más especificaciones

Descripción

Descripción

  • Sequence: MSPCPPQQSRNRVIQLSTSELGEMELTWQEIMSITELQGLNAPSEPSFEPQAPAPYLGPPPPTTYCPCSIHPDSGFPLPPPPYELPASTSHVPDPPYSYGNMAIPVSKPLSLSGLLSEPLQDPLALLDIGLPAGPPKPQEDPESDSGLSLNYSDAESLELEGTEAGRRRSEYVEMYPVEYPYSLMPNSLAHSNYTLPAAETPLALEPSSGPVRAKPTARGEAGSRDERRALAMKIPFPTDKIVNLPVDDFNELLARYPLTESQLALVRDIRRRGKNKVAAQNCRKRKLETIVQLERELERLTNERERLLRARGEADRTLEVMRQQLTELYRDIFQHLRDESGNSYSPEEYALQQAADGTIFLVPRGTKMEATD
Especificaciones

Especificaciones

Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4778
Videos
SDS
Documentos

Documentos

Product Certifications