missing translation for 'onlineSavingsMsg'
Learn More

Neurogranin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10440-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816833

  • 319.00€ / 150 microlitros

Envío estimado: 20-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.
Especificaciones

Especificaciones

Neurogranin
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P60761, Q04940, Q92686
hng, Neurogranin, Ng, NRGN, P17, RC3
Rabbit
IgG
150 μL
-20°C
Primary
4900, 64011, 64356
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
NRGN
Liquid
NRGN
Neurogranin Fusion Protein Ag0676
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados