missing translation for 'onlineSavingsMsg'
Learn More

Sulfato de neodimio(III) octahidrato, REacton™, 99,99 % (REO), Thermo Scientific Chemicals

Catalog Number p-7049195
Click to view available options
Cantidad:
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
13477-91-3
MFCD00149803
Este artículo no se puede devolver. Vea la política de devoluciones
13477-91-3
MFCD00149803

Neodymium(III) sulfate is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Neodymium(III) sulfate octahydrate
Punto de fusión 1176°C
Forma física cristalino/a/s
Cantidad 25 g
Notas del porcentaje del ensayo (REO)
Índice Merck 14,6450
Información de solubilidad Soluble in water (80g/L) at 20°C moderately soluble in strong mineral acids.
Formula Weight (peso de la fórmula) 720.78 (576.66 Anhydrous)
Porcentaje de pureza 99.99%
Grado Reacton™
Olor Odorless
Densidad 2.85 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'