missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfato de neodimio(III) octahidrato, REacton™, 99,99 % (REO), Thermo Scientific Chemicals
Click to view available options
Cantidad:
25 g
100 g
Descripción
Neodymium(III) sulfate is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Neodymium(III) sulfate octahydrate |
Punto de fusión | 1176°C |
Forma física | Crystalline |
Cantidad | 100 g |
Notas del porcentaje del ensayo | (REO) |
Índice Merck | 14,6450 |
Formula Weight (peso de la fórmula) | 720.78 (576.66 Anhydrous) |
Porcentaje de pureza | 99.99% |
Grado | Reacton™ |
Olor | Odorless |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'