missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK7 Recombinant Human Protein
NEK7 Recombinant Human Protein
Marca: Invitrogen PV3833
Detalles adicionales : Peso : 0.05250kg
Descripción
NEK7 belongs to the NIMA family of kinases which are involved in DNA replication and genotoxic stress response.Especificaciones
NP_598001.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
63.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
NEK7 | |
NEK7 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 667μg/mL NEK6 peptide substrate (FLAKSFGSPNRAYKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 8-313 | |
3nmoles of phosphate transferred to NEK6 peptide substrate (FLAKSFGSPNRAYKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL | |
GST Tag | |
Kinase and inhibitors | |
80% by SDS-PAGE |
0.34mg/mL | |
140609, 4086 | |
NEK7 Recombinant Human Protein | |
–80°C | |
1 Each | |
NEK7 Recombinant Human Protein | |
NEK | |
NEK7 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.