missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK4 Recombinant Human Protein
NEK4 is a serine/threonine kinase.
Marca: Invitrogen™ PV4331
Detalles adicionales : Peso : 0.00900kg
Descripción
NEK4 is involved in signal transduction and cellular communication. It is proposed to act exclusively upon threonine residues and is present in most primary carcinomas.
Especificaciones
NP_003148 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
NEK4, NRK2 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
122kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |