missing translation for 'onlineSavingsMsg'
Learn More

Nectin-2/CD112 Antibody (005), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89188IR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213938

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

Nectin-2/CD112
Monoclonal
DyLight 755
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Mouse PVRL2
Store at 4°C in the dark.
IgG
Flow Cytometry
5
Flow Cytometry
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
This antibody was obtained from a rabbit immunized with purified, recombinant Human Nectin-2/CD112 extracellular domain (Met 1-Leu 360; NP_002847.1).
0.1 mL
Immunology
5819
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales