Learn More
Nebivolol Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Nebivolol Hydrochloride |
| CAS | 152520-56-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H25 F2 N O4 . Cl H |
| Quantity | 10 mg |
| Synonym | (αR,α'R,2R,2'S)-rel-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride (1:1), (αR,α'R,2R,2'S)-rel-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride, [2R*[R*[R*(S*)]]]-(±)-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol hydrochloride, Nebicard, Nebilet, Nebivolol hydrochloride, Nodon, R 067555 |
| InChI Formula | InChI=1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m1./s1 |
| SMILES | Cl.O[C@H](CNC[C@@H](O)[C@H]1CCc2cc(F)ccc2O1)[C@@H]3CCc4cc(F)ccc4O3 |
| IUPAC Name | (1R)-1-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-[[(2R)-2-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]ethanol;hydrochloride |
| Molecular Weight (g/mol) | 441.9 |
| Show More |