missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17614-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It plays a important role in transferring electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. Hydropathy analysis revealed that this subunit and 4 other subunits have an overall hydrophilic pattern, even though they are found within the hydrophobic protein (HP) fraction of complex I.Especificaciones
NDUFB2 | |
Polyclonal | |
Unconjugated | |
NDUFB2 | |
AGGG, CI AGGG, Complex I AGGG, NDUFB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362344, 4708, 68198 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95178, Q9CPU2 | |
Ndufb2 | |
NDUFB2 Fusion Protein Ag11789 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |