missing translation for 'onlineSavingsMsg'
Learn More

NDNL2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 27488-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16815655

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is part of the SMC5-6 chromatin reorganizing complex and is a member of the MAGE superfamily. This is an intronless gene. Component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintece via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs).
Especificaciones

Especificaciones

NDNL2
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96MG7
HCA4, MAGE G1 antigen, MAGEG1, MAGEL3, Melanoma associated antigen G1, NDNL2, necdin like 2, Necdin like protein 2, NSE3, NSMCE3
Rabbit
IgG
150 μL
-20°C
Primary
56160
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NSMCE3
Liquid
NSMCE3
NDNL2 Fusion Protein Ag26663
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados