Learn More
Ansell Edmont™ Marigold™ Natural Rubber Industrial Protective Glove
Catalog Number p-8010802
Click to view available options
Tamaño:
7.5
8.5
10.5
Ideal for use with heavy duty protection.
  • Chemical
  • Unlined black natural rubber latex glove
  • Chlorinated to harden and cleanse the surface of the glove
  • Beaded cuff for tear resistance and easy donning
Color Black
Acabado exterior Smooth
Material Rubber Latex
Tamaño 8.5
Estilo de puño de los guantes Beaded
Resistente a Chemicals