Learn More
Napsin A Antibody (TMU-Ad02), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB110-68133AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214023
  • 572.34€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
Napsin A
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
Mouse
Protein A purified
RUO
Primary
Protein A purified
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TMU-Ad02
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
O96009
NAPSA
Synthetic peptide corresponding to the N-terminus of human NAPSA [UniProt# O96009]
0.1 mL
Cancer, Cell Biology, Cellular Markers
9476
Human
Purified