Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Product Code. 17124791
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
300 kDa
Includes:
100/Pk.
24/Pk.
500/Pk.
Color:
Blue
Clear
Gray
Orange
Red
Unit Size:
Pack of 100
Pack of 24
Pack of 500
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Includes Color unitSize
17124791 100 kDa 100/Pk. Clear Pack of 100
17164791 300 kDa 100/Pk. Orange Pack of 100
17154791 300 kDa 24/Pk. Orange Pack of 24
17114791 100 kDa 24/Pk. Clear Pack of 24
17184781 30 kDa 100/Pk. Red Pack of 100
17174781 30 kDa 24/Pk. Red Pack of 24
17134781 10 kDa 24/Pk. Blue Pack of 24
17104781 3 kDa 100/Pk. Gray Pack of 100
17194771 3 kDa 24/Pk. Gray Pack of 24
17144781 10 kDa 100/Pk. Blue Pack of 100
17154781 10 kDa 500/Pk. Blue Pack of 500
17114781 3 kDa 500/Pk. Gray Pack of 500
17194781 30 kDa 500/Pk. Red Pack of 500
17134791 100 kDa 500/Pk. Clear Pack of 500
17174791 300 kDa 500/Pk. Orange Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 17124791 Supplier Cytiva Supplier No. OD100C34
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Description EFA: 0.3 cm2
Material (Membrane) PES
MWCO 100 kDa
Sterility Non-sterile
Housing Material Polypropylene
Includes 100/Pk.
Volume (Metric) 50 to 500 μL (Sample)