Learn More
Naftifine Hydrochloride, TRC
Product Code. 30134031
Change view
Click to view available options
Quantity:
50 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134031 500 mg 500mg
30138300 2.5 g 2.5g
30012029 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134031 Supplier Toronto Research Chemicals Supplier No. TRCN213100500MG
 Restricted Item
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 65473-14-5
Molecular Formula C21 H21 N . Cl H
Molecular Weight (g/mol) 323.86
Synonym N-Methyl-N-[(2E)-3-phenyl-2-propen-1-yl]-1-Naphthalenemethanamine Hydrochloride (1:1), 1-Naphthalenemethanamine, N-methyl-N-(3-phenyl-2-propenyl)-, hydrochloride, (E)-, 1-Naphthalenemethanamine, N-methyl-N-[(2E)-3-phenyl-2-propenyl]-, hydrochloride (9CI), AW 105-843, Exoderil, Naftifine hydrochloride, Naftin, SN 105-843
IUPAC Name (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine;hydrochloride
SMILES Cl.CN(C\C=C\c1ccccc1)Cc2cccc3ccccc23
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C21H21N.ClH/c1-22(16-8-11-18-9-3-2-4-10-18)17-20-14-7-13-19-12-5-6-15-21(19)20;/h2-15H,16-17H2,1H3;1H/b11-8+;
Formula Weight 323.1441
Chemical Name or Material Naftifine Hydrochloride