missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NA-XTD™ Influenza Neuraminidase Assay Kit

Next-generation chemiluminescence-based assay for long-lived emission kinetics for quantitation of influenza neuraminidase inhibitor resistance. Thermo Scientific NA-XTD Influenza Neuraminidase Assay Kit can be used for virus quantitation in media samples

1406.00€ - 1452.00€

Especificaciones

Para utilizar con (aplicación) Assessing and researching drug resistance of influenza. Screening for new neuraminidase inhibitors and quantitating viral neuraminidase activity for vaccine development
Formato 96-well plate
Requisitos de almacenamiento Store all components at 2° to 8°C.
Ver más especificaciones

Productos 2
Código de producto Marca Tipo de producto Precio Cantidad y disponibilidad  
Código de producto Marca Tipo de producto Precio Cantidad y disponibilidad  
12604265 Ver documentos Invitrogen™
4457534
NA-XTD Influenza Neuraminidase Assay Reagent Set 1406.00€
cada uno Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 07-10-2019
Añadir a la cesta
 
12614265 Ver documentos Invitrogen™
4457535
NA-XTD Influenza Neuraminidase Assay Kit 1452.00€
cada uno Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 07-10-2019
Añadir a la cesta
 
Descripción

Descripción

The NA-XTD kit includes detection reagents and microplates, eliminating the need for luminometers equipped with a reagent injector, and improving ease-of-use. This kit also includes complete assay protocols for quantitating sensitivity to neuraminidase inhibitors in various influenza virus isolates including: human influenza types A and B, A/H1N1 pandemic, avian, equine and porcine viruses.
 
  • Superior performance - improved, long-lived light emission kinetics and read-time flexibility
  • High sensitivity - high assay signal-to-noise, detection of low-virus concentrations, and wide assay dynamic range
  • Simple instrumentation requirement - no reagent injectors or luminometer needed
  • Multiple applications - neuraminidase inhibitor IC50 assays and cell-based virus quantitation
Long-Lived Light Emission Kinetics and Read-Time Flexibility
The NA-XTD assay yields much longer-lived light emission kinetics than does the NA-Star assay, eliminating the need for luminometer instrumentation with reagent injectors and enabling read-time flexibility and batch-mode processing of assay plates. NA-XTD assay signal half-life is approximately 2 hours, longer than the 5-10 minute half-life of the NA-Star assay signal. IC-50 values determined from data collected immediately or up to 3 hours following addition of NA-XTD Accelerator solution are identical.
 
High Sensitivity
The NA-XTD chemiluminescent assay provides higher detection sensitivity (better low-end detection limit), higher assay signal-to-noise ratio, and wider assay dynamic range than fluorescent assays with the MUNANA substrate. The NA-XTD assay also typically demonstrates slightly higher signal-to-noise than the NA-Star assay. The NA-XTD assay provides 2 to 50-fold higher sensitivity by signal-to-noise ratio than MUNANA-based fluorescence assays, depending on the virus isolate. The NA-XTD assay provides a dynamic range of detection of 3 to 4 orders of magnitude of neuraminidase enzyme concentration, compared to 2 to 3 orders of magnitude range with fluorescent MUNANA assays. The wide chemiluminescent neuraminidase assay range enables determination of IC-50 values over a wide range of virus concentrations, eliminating the need to titer virus prior to performing IC50 determination assays.
 
Simple Instrumentation Requirement
Optimally, a small sample of culture media is removed and assayed directly with NA-XTD reagents, permitting multiple samples to be assayed over time.
 
Specifications
  • Detection Method: Chemiluminescent
  • For Use With Equipment: Luminometer (Microplate)
  • Form: Liquid
  • Format: 96-well plate
  • High Throughput Compatibility: High Throughput-Compatible
  • Readout: End Point
  • Substrate: NA-XTD
  • Substrate Properties: Chemical Substrate
  • Substrate Type: Neuraminidase Substrate
  • Target Enzyme: Neuraminidase, Sialidase
Especificaciones

Especificaciones

Assessing and researching drug resistance of influenza. Screening for new neuraminidase inhibitors and quantitating viral neuraminidase activity for vaccine development
96-well plate
Store all components at 2° to 8°C.

For Research Use Only. Not for use in diagnostics procedures.