Learn More
N-(tert-Butyloxy)carbonyl-L-thyroxine, TRC
Product Code. 30348727
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
Product Code. Quantity Unit Size
30348727 2.5 g 2.50g
1 options
This item is not returnable. View return policy
Product Code. 30348727 Supplier Toronto Research Chemicals Supplier No. TRCB6937052.5G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(tert-Butyloxy)carbonyl-L-thyroxine
CAS 88404-22-2
Molecular Formula C20H19I4NO6
Quantity 2.5 g
Synonym N-[(1,1-Dimethylethoxy)carbonyl]-O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine
InChI Formula InChI=1S/C20H19I4NO6/c1-20(2,3)31-19(29)25-15(18(27)28)6-9-4-13(23)17(14(24)5-9)30-10-7-11(21)16(26)12(22)8-10/h4-5,7-8,15,26H,6H2,1-3H3,(H,25,29)(H,27,28)/t15-/m0/s1
SMILES IC(C=C(C[C@@H](C(O)=O)NC(OC(C)(C)C)=O)C=C1I)=C1OC2=CC(I)=C(O)C(I)=C2
IUPAC Name (S)-2-((tert-butoxycarbonyl)amino)-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
Molecular Weight (g/mol) 876.99
Formula Weight 876.74