missing translation for 'onlineSavingsMsg'
Learn More

N-[(S)-alfa-(4-Clorofenil)bencil]formamida, 98 %, Thermo Scientific™

Catalog Number p-7053755
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
474654-19-8
C14H12ClNO
245.71
HDGOSEHHSVUCBO-UHFFFAOYNA-N
n-s-4-chlorophenyl phenyl methyl formamide, n-s-4-chlorophenyl phenylmethyl-formamide
Este artículo no se puede devolver. Vea la política de devoluciones
474654-19-8
C14H12ClNO
245.71
HDGOSEHHSVUCBO-UHFFFAOYNA-N
n-s-4-chlorophenyl phenyl methyl formamide, n-s-4-chlorophenyl phenylmethyl-formamide

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 245.7
Porcentaje de pureza 98%
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'