missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl formate, 97%

Product Code. 11469236
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
11469236 500 mL 500mL
11459236 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 11469236

Brand: Thermo Scientific Alfa Aesar A18090.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-74-7 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-74-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00003295
InChI Key KFNNIILCVOLYIR-UHFFFAOYSA-N
Synonym formic acid, propyl ester, n-propyl formate, propyl methanoate, formiate de propyle, formic acid propyl ester, propylester kyseliny mravenci, unii-wo1arv6gtw, formiate de propyle french, fema no. 2943, wo1arv6gtw
PubChem CID 8073
IUPAC Name propyl formate
SMILES CCCOC=O

Specifications

Melting Point -93°C
Density 0.904
Boiling Point 80°C to 81°C
Flash Point −4°C (25°F)
Refractive Index 1.375
Quantity 500 mL
UN Number UN1281
Beilstein 1739272
Merck Index 14,7858
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material n-Propyl formate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.