Learn More
Acetal dibencílico de N,N-dimetilformamida, téc. 80 %, Thermo Scientific Chemicals
Catalog Number p-7081320
Click to view available options
Cantidad:
10 g
50 g
250 g
2016-04-8
C17H22NO2
272.37
MFCD00014869
JFIKHFNGAURIIB-UHFFFAOYSA-O
n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
74837
N,N-dimetil-1,1-bis(fenilmetoxi)metanamina
CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2
2016-04-8
C17H22NO2
272.37
MFCD00014869
JFIKHFNGAURIIB-UHFFFAOYSA-O
n,n-dimethylformamide dibenzyl acetal, n,n-dimethylformamide dibenzylacetal, 1,1-bis benzyloxy trimethylamine, methanamine, n,n-dimethyl-1,1-bis phenylmethoxy, bis benzyloxy methyl dimethylamine, acmc-1cro2, 1,1-dibenzyloxytrimethylamine, dimethyl formamide dibenzylacetal, bis benzyloxy methyldimethylamine, dimethyl formamide dibenzyl acetal
74837
N,N-dimetil-1,1-bis(fenilmetoxi)metanamina
CN(C)C(OCC1=CC=CC=C1)OCC2=CC=CC=C2
N,N-Dimethylformamide diethyl acetal was used in quantification of cocaine and its primary metabolite, benzoyl ecgonine from urine matrix by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El acetal dietílico de N,N-dimetilformamida se utilizó en la cuantificación de la cocaína y su metabolito primario, la bencilecgonina, en la matriz de orina mediante cromatografía de gases.
Solubilidad
Soluble en diversos disolventes inertes.
Notas
Almacenar alejado de agentes oxidantes fuertes. Mantener el recipiente herméticamente cerrado. Almacenar en un lugar fresco y seco y en recipientes bien sellados.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N,N-Dimethylformamide dibenzyl acetal
Densidad 1.045
Punto de ebullición 138°C to 140°C (0.5 mmHg)
Cantidad 10 g
Sensibilidad Moisture sensitive
Información de solubilidad Soluble in variety of inert solvents.
Formula Weight (peso de la fórmula) 271.36
Porcentaje de pureza 85%
Grado Técnico