Learn More
Alfa Aesar™ N,N-Dimethylcyanoacetamide, 98%
Marca: Alfa Aesar™ A10981.06
Detalles adicionales : CAS : 7391-40-4 Peso : 0.00500kg
Cantidad. | 5g |
---|
Identificadores de productos químicos
7391-40-4 | |
112.132 | |
MATJPVGBSAQWAC-UHFFFAOYSA-N | |
251143 | |
CN(C)C(=O)CC#N |
C5H8N2O | |
MFCD00051623 | |
n,n-dimethylcyanoacetamide, acetamide, 2-cyano-n,n-dimethyl, n,n-dimethyl cyanoacetamide, n,n-dimethyl-2-cyanoacetamide, acmc-209osw, n,n-dimethylcyanoacetate, 2-cyano-n,n-dimethyl-acetamide, 2-cyano-n,n-dimethyl-ethanamide, acetamide,2-cyano-n,n-dimethyl, cyanoacetic acid n,n-dimethylamide | |
2-cyano-N,N-dimethylacetamide |
Especificación
N,N-Dimethylcyanoacetamide | |
5g | |
C5H8N2O | |
773950 | |
MATJPVGBSAQWAC-UHFFFAOYSA-N | |
2-cyano-N,N-dimethylacetamide | |
251143 | |
98% |
59°C to 66°C | |
7391-40-4 | |
MFCD00051623 | |
n,n-dimethylcyanoacetamide, acetamide, 2-cyano-n,n-dimethyl, n,n-dimethyl cyanoacetamide, n,n-dimethyl-2-cyanoacetamide, acmc-209osw, n,n-dimethylcyanoacetate, 2-cyano-n,n-dimethyl-acetamide, 2-cyano-n,n-dimethyl-ethanamide, acetamide,2-cyano-n,n-dimethyl, cyanoacetic acid n,n-dimethylamide | |
CN(C)C(=O)CC#N | |
112.132 | |
112.13 |