missing translation for 'onlineSavingsMsg'
Learn More

Ácido n-metiliminodiacético, 99 %, Thermo Scientific Chemicals

Catalog Number p-164464
Click to view available options
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Este artículo no se puede devolver. Vea la política de devoluciones
4408-64-4
C5H9NO4
147.13
MFCD00004284
XWSGEVNYFYKXCP-UHFFFAOYSA-N
n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
20441
ácido 2-[carboximetil(metil)amino]acético
CN(CC(=O)O)CC(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
4408-64-4
C5H9NO4
147.13
MFCD00004284
XWSGEVNYFYKXCP-UHFFFAOYSA-N
n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
20441
ácido 2-[carboximetil(metil)amino]acético
CN(CC(=O)O)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Methyliminodiacetic acid
Nombre de nota 99%
% mín. CAS 98.5
% máx. CAS 100.0
Punto de fusión 218°C to 221°C
Color Crema a blanco
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 99%
Envase Frasco de vidrio
Fórmula lineal CH3N(CH2CO2H)2
Cantidad 5 g
Beilstein 04, 367
Formula Weight (peso de la fórmula) 147.13
Porcentaje de pureza 99%
Forma física Polvo
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'