missing translation for 'onlineSavingsMsg'
Learn More

N-Metil-N-[2-nitro-4-(trifluorometil)fenil]hidracina, 97 %, Alfa Aesar™

Catalog Number p-7027798
Click to view available options
Cantidad:
0.25 g
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
120940-43-4
C8H8F3N3O2
235.166
MFCD00052691
GBBAZCATUSUQOJ-UHFFFAOYSA-N
1-methyl-1-2-nitro-4-trifluoromethyl phenyl hydrazine, n-methyl-n-2-nitro-4-trifluoromethyl phenyl hydrazine, hydrazine,1-methyl-1-2-nitro-4-trifluoromethyl phenyl, aminomethyl 2-nitro-4-trifluoromethyl phenyl amine, acmc-1cirx, 1-methyl-1-2-nitro-4-trifluoromethyl phenyl diazane
2775458
1-metil-1-[2-nitro-4-(trifluorometil)fenil]hidracina
CN(C1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-])N
Este artículo no se puede devolver. Vea la política de devoluciones
120940-43-4
C8H8F3N3O2
235.166
MFCD00052691
GBBAZCATUSUQOJ-UHFFFAOYSA-N
1-methyl-1-2-nitro-4-trifluoromethyl phenyl hydrazine, n-methyl-n-2-nitro-4-trifluoromethyl phenyl hydrazine, hydrazine,1-methyl-1-2-nitro-4-trifluoromethyl phenyl, aminomethyl 2-nitro-4-trifluoromethyl phenyl amine, acmc-1cirx, 1-methyl-1-2-nitro-4-trifluoromethyl phenyl diazane
2775458
1-metil-1-[2-nitro-4-(trifluorometil)fenil]hidracina
CN(C1=C(C=C(C=C1)C(F)(F)F)[N+](=O)[O-])N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Methyl-N-[2-nitro-4-(trifluoromethyl)phenyl]hydrazine
Punto de fusión 64°C to 67°C
Cantidad 0.25 g
Formula Weight (peso de la fórmula) 235.16
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'