missing translation for 'onlineSavingsMsg'
Learn More

N-metil-D-fenilalanina, 97 %, Thermo Scientific™

Catalog Number p-7056639
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
56564-52-4
C10H13NO2
179.219
MFCD00069560
SCIFESDRCALIIM-SECBINFHSA-N
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
6427037
Ácido (2R)-2-(metilamino)-3-fenilpropanoico
CNC(CC1=CC=CC=C1)C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
56564-52-4
C10H13NO2
179.219
MFCD00069560
SCIFESDRCALIIM-SECBINFHSA-N
r-2-methylamino-3-phenylpropanoic acid, n-methyl-d-phenylalanine, d-phenylalanine, n-methyl, 2r-2-methylamino-3-phenylpropanoic acid, h-d-mephe-oh hcl, ambotzhaa1199, l-phenylalanine, n-methyl
6427037
Ácido (2R)-2-(metilamino)-3-fenilpropanoico
CNC(CC1=CC=CC=C1)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Methyl-D-phenylalanine
Beilstein 3199933
Formula Weight (peso de la fórmula) 179.22
Porcentaje de pureza 97%
Cantidad 250 mg
Forma física Sólido
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'