Learn More
N-Methyl-(6-thien-2-ylpyrid-3-yl)methylamine, 97%, Thermo Scientific™
Product Code. 10623825
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10623825 250 mg Amber Glass Bottle 250mg
10225424 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623825 Supplier Thermo Scientific Maybridge Supplier No. CC54846CB
 Restricted Item
Fecha estimada de envoi: 04-12-2026
Add to basket
This item is not returnable. View return policy
Chemical Identifiers
CAS 886851-41-8
Molecular Formula C11H12N2S
Molecular Weight (g/mol) 204.291
MDL Number MFCD09064971
InChI Key LFZDJRPWILOVEO-UHFFFAOYSA-N
Synonym n-methyl-6-thien-2-ylpyrid-3-yl methylamine, n-methyl-1-6-thien-2-yl pyridin-3-yl methylamine, 3-pyridinemethanamine,n-methyl-6-2-thienyl, methyl 6-thiophen-2-yl pyridin-3-yl methyl amine, n-methyl-1-6-thiophen-2-yl pyridin-3-yl methanamine, methyl 6-2-thienyl 3-pyridyl methyl amine
PubChem CID 24229544
IUPAC Name N-methyl-1-(6-thiophen-2-ylpyridin-3-yl)methanamine
SMILES CNCC1=CN=C(C=C1)C2=CC=CS2
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.3
Percent Purity 97%
Chemical Name or Material N-methyl-(6-thien-2-ylpyrid-3-yl)methylamine