missing translation for 'onlineSavingsMsg'
Learn More

N-Metil-2-bifenilmetilamina, 99 %, Thermo Scientific™

Catalog Number p-7033622
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
13737-31-0
C14H15N
197.281
MFCD00598943
FXXBKXHFEBSUHS-UHFFFAOYSA-N
n-methyl-2-biphenylmethylamine, 1-1,1'-biphenyl-2-yl-n-methylmethanamine, methyl 2-phenylphenyl methyl amine, 1-biphenyl-2-yl-n-methylmethylamine, 1,1'-biphenyl-2-ylmethyl methyl amine, acmc-20aoxt, n-methyl-2-phenylbenzylamine, 1-biphenyl-2-yl-methylmethylamine, 1-biphenyl-2-yl-n-methylmethanamine, 1,1'-biphenyl-2-methanamine,n-methyl
19366385
N-metil-1-(2-fenilfenil)metanamina
CNCC1=CC=CC=C1C2=CC=CC=C2
Este artículo no se puede devolver. Vea la política de devoluciones
13737-31-0
C14H15N
197.281
MFCD00598943
FXXBKXHFEBSUHS-UHFFFAOYSA-N
n-methyl-2-biphenylmethylamine, 1-1,1'-biphenyl-2-yl-n-methylmethanamine, methyl 2-phenylphenyl methyl amine, 1-biphenyl-2-yl-n-methylmethylamine, 1,1'-biphenyl-2-ylmethyl methyl amine, acmc-20aoxt, n-methyl-2-phenylbenzylamine, 1-biphenyl-2-yl-methylmethylamine, 1-biphenyl-2-yl-n-methylmethanamine, 1,1'-biphenyl-2-methanamine,n-methyl
19366385
N-metil-1-(2-fenilfenil)metanamina
CNCC1=CC=CC=C1C2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Methyl-2-biphenylmethylamine
Cantidad 250 mg
Formula Weight (peso de la fórmula) 197.28
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'