missing translation for 'onlineSavingsMsg'
Learn More

N-(hidroximetil)ftalimida, 97 %, Thermo Scientific Chemicals

Catalog Number 10245400
Click to view available options
Cantidad:
25 g
100 g
Envase:
Frasco de vidrio
Este artículo no se puede devolver. Vea la política de devoluciones
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
8354
CHEBI:38816
2-(hidroximetil)isoindol-1,3-diona
C1=CC=C2C(=C1)C(=O)N(C2=O)CO
Este artículo no se puede devolver. Vea la política de devoluciones
118-29-6
C9H7NO3
177.16
MFCD00005899
MNSGOOCAMMSKGI-UHFFFAOYSA-N
n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
8354
CHEBI:38816
2-(hidroximetil)isoindol-1,3-diona
C1=CC=C2C(=C1)C(=O)N(C2=O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-(Hydroxymethyl)phthalimide
Nombre de nota 97%
% mín. CAS 96.0
% máx. CAS 100.0
Punto de fusión 140°C to 145°C
Color Blanco
Pérdida por secado 2% max.
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 96% min. (HPLC)
Envase Frasco de vidrio
Cantidad 100 g
Beilstein 21, 475
Fieser 01,484; 08,253
Formula Weight (peso de la fórmula) 177.16
Porcentaje de pureza 97%
Forma física Polvo cristalino fino
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'