Learn More
Acrilato de n-hexilo, 95 %, estabilizado con hidroquinona, Thermo Scientific Chemicals
Catalog Number p-7045237
Click to view available options
Cantidad:
25 g
100 g
2499-95-8
C9H16O2
156.225
MFCD00048881
LNMQRPPRQDGUDR-UHFFFAOYSA-N
hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
17259
prop-2-enoato de hexilo
CCCCCCOC(=O)C=C
2499-95-8
C9H16O2
156.225
MFCD00048881
LNMQRPPRQDGUDR-UHFFFAOYSA-N
hexyl acrylate, n-hexyl acrylate, hexyl 2-propenoate, 2-propenoic acid, hexyl ester, n-hexylacrylate, ageflex n-ha, acrylic acid, hexyl ester, ccris 7038, acrylic acid-n-hexyl ester, hexylacrylate
17259
prop-2-enoato de hexilo
CCCCCCOC(=O)C=C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material n-Hexyl acrylate
Nombre de nota stabilized with hydroquinone
Punto de fusión -45°C
Densidad 0.89
Punto de ebullición 88°C to 89°C (24 mmHg)
Temperatura de inflamación 68°C (154°F)
Fórmula lineal H2C=CHCO2(CH2)5CH3
Índice de refracción 1.428
Cantidad 25 g
Número UN UN3082
Beilstein 1757327
Formula Weight (peso de la fórmula) 156.22
Porcentaje de pureza 95%