Learn More
N-Ethyl-4-methylbenzenesulfonamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30310885
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
30310885 1 mL 1mL
1 options
This item is not returnable. View return policy
Product Code. 30310885 Supplier Dr. Ehrenstorfer Supplier No. DREA13348005AL100
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Ethyl-4-methylbenzenesulfonamide
CAS 80-39-7
Analyte or Component Names N-Ethyl-4-methylbenzenesulfonamide
Shipping Condition +20°C
Molecular Formula C9 H13 N O2 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym p-Toluenesulfonamide, N-ethyl- (6CI,7CI,8CI), N-Ethyl-4-methylbenzenesulfonamide, N-Ethyl-4-toluenesulfonamide, N-Ethyl-p-methylbenzenesulfonamide, N-Ethyl-p-toluenesulfonamide, N-Ethyl-p-tolylsulfonamide, N-Ethyltosylamide, N-Tosylethylamine, NSC 68803, Santicizer 3, p-Toluene-N-ethylsulfonamide, p-Toluenesulfonyl-N-ethylamide
InChI Formula InChI=1S/C9H13NO2S/c1-3-10-13(11,12)9-6-4-8(2)5-7-9/h4-7,10H,3H2,1-2H3
SMILES CCNS(=O)(=O)c1ccc(C)cc1
IUPAC Name N-ethyl-4-methylbenzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 199.27
Quantity 1 mL
Formula Weight 199.0667 g/mol