Learn More
N-Desmethyl Doxycycline, TRC
Product Code. 30365358
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365358 50 mg 50mg
30364660 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365358 Supplier Toronto Research Chemicals Supplier No. TRCD29242550MG
 Restricted Item
Fecha estimada de envoi: 16-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86271-83-2
Molecular Formula C21H22N2O8
Molecular Weight (g/mol) 430.41
Synonym [4S-(4alpha,4aalpha,5alpha,5aalpha,6alpha,12aalpha)]-1,4,4a,5,5a,6,11,12a-Octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-4-(methylamino)-1,11-dioxo-2-naphthacenecarboxamide
SMILES CN[C@H]1[C@@H]2[C@@H](O)[C@@H]3[C@@H](C)c4cccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C21H22N2O8/c1-6-7-4-3-5-8(24)10(7)15(25)11-9(6)16(26)13-14(23-2)17(27)12(20(22)30)19(29)21(13,31)18(11)28/h3-6,9,13-14,16,23-26,29,31H,1-2H3,(H2,22,30)/t6-,9+,13+,14-,16-,21-/m0/s1
Formula Weight 430.1376
Chemical Name or Material N-Desmethyl Doxycycline