missing translation for 'onlineSavingsMsg'
Learn More

N-Ciclohexil-N-metil-4-nitrobenzamida, 97 %, Thermo Scientific™

Catalog Number p-7040597
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
29477-27-8
C14H18N2O3
262.309
MFCD00709770
ZGDQZPUGCAXWLR-UHFFFAOYSA-N
benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
692212
N-ciclohexil-N-metil-4-nitrobenzamida
CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
29477-27-8
C14H18N2O3
262.309
MFCD00709770
ZGDQZPUGCAXWLR-UHFFFAOYSA-N
benzamide, n-cyclohexyl-n-methyl-4-nitro, aronis24032, n-cyclohexyl-n-methyl 4-nitrophenyl carboxamide
692212
N-ciclohexil-N-metil-4-nitrobenzamida
CN(C1CCCCC1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material N-Cyclohexyl-N-methyl-4-nitrobenzamide
Cantidad 250 mg
Formula Weight (peso de la fórmula) 262.3
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'